Anti-PTPRC Mouse Polyclonal Antibody [clone: JS-83]
ORIGTA320452
: OriGene
- Antibody type:Primary
- Antigen name:Protein tyrosine phosphatase receptor type C
- Antigen symbol:PTPRC
- Clonality:Polyclonal
- Clone:JS-83
- Conjugation:Unconjugated
- Host:Mouse
- Reactivity:Human
- Cross adsorption:No
- Form:Liquid
- Storage buffer:Aqueous buffer, 0,09% sodium azide, may contain carrier protein/stabilizer
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Purification:purified by affinity chromatography
- Size:100 μg
- Pk:100 µG
Type: Primary
Antigen: PTPRC
Clonality: Polyclonal
Clone: JS-83
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human