Anti-PTPRC Rat Polyclonal Antibody [clone: 30-F11]
Catalog # ORIGTA320448
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen name:Protein tyrosine phosphatase receptor type C
- Antigen symbol:PTPRC
- Clonality:Polyclonal
- Clone:30-F11
- Conjugation:Unconjugated
- Host:Rat
- Reactivity:Mouse
- Cross adsorption:No
- Form:Liquid
- Storage buffer:Aqueous buffer, 0,09% sodium azide, may contain carrier protein/stabilizer
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Purification:purified by affinity chromatography
- Size:100 μg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: PTPRC
Clonality: Polyclonal
Clone: 30-F11
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype:
Reactivity: Mouse