Anti-KIR2DL3 Rabbit Polyclonal Antibody
Catalog # ORIGTA321841
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen symbol:KIR2DL3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Form:Liquid
- Amino acid number:corresponding to a region between amino acids 22 to 245
- Storage buffer:PBS pH 7,3, 0,05% NaN3, 50% glycerol
- Molecular weight:38 kDa
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:Fusion protein corresponding to a region derived from 22-245 amino acids of human killer cell immunoglobulin-like receptor, two domains, long cytoplasmic tail, 3
- Purification:purified by antigen affinity chromatography
- Size:100 μl
- Pk:100 µl
Specifications
About this item
Recommended Dilutions: Western Blot: 1:500-2000
Type: Primary
Antigen: KIR2DL3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human