Order Entry
Czech Republic
ContactUsLinkComponent
 
Anti-PTAFR Rabbit Polyclonal Antibody
Catalog # ORIGTA321248
Supplier:  OriGene
Anti-PTAFR Rabbit Polyclonal Antibody
Catalog # ORIGTA321248
Supplier:  OriGene

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    PTAFR
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Cross adsorption:
    No
  • Form:
    Liquid
  • Amino acid number:
    1 to 14
  • Storage buffer:
    PBS pH 7,3, 0,05% NaN3, 50% glycerol
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    –20 °C
  • Immunogen:
    Synthetic peptide corresponding to a region derived from 1-14 amino acids of Human platelet-activating factor receptor
  • Purification:
    purified by antigen affinity chromatography
  • Size:
    100 μl
  • Pk:
    100 µl

Specifications

About this item

Recommended Dilutions: ELISA: 1:1000-5000; Immunochemistry: 1:25-100

Type: Primary
Antigen: PTAFR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat