Anti-ITGB4 Rat Polyclonal Antibody [clone: 439-9B]
Catalog # ORIGTA320398
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen symbol:ITGB4
- Clonality:Polyclonal
- Clone:439-9B
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Rat
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Form:Liquid
- Storage buffer:Aqueous buffer, 0,09% sodium azide, may contain carrier protein/stabilizer
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Purification:purified by affinity chromatography
- Size:100 μg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: ITGB4
Clonality: Polyclonal
Clone: 439-9B
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype:
Reactivity: Human