Anti-SOX8 Rabbit Polyclonal Antibody
Catalog # ORIGTA322372
Supplier: OriGene
Specifications
- Druh protilátky:Primární
- Symbol antigenu:SOX8
- Klonovalita:Polyclonal
- Spojení:Unconjugated
- Hostitel:Rabbit
- Izotyp:IgG
- Reaktivita:Human,Rat,Mouse
- Western blot:Yes
- Křížová adsorbce:No
- Formulář:Liquid
- Počet aminokyselin:407 to 419
- Skladovací pufr:PBS pH 7,3, 0,05% NaN3, 50% glycerol
- Molekulární hmotnost:47 kDa
- Skladovací teplota:–20 °C
- Transportní teplota:–20 °C
- Imunogen:Synthetic peptide corresponding to a region derived from 407-419 amino acids of human SRY (sex determining region Y)-box 8
- Čištění:purified by antigen affinity chromatography
- Rozměry protilátky:100 μl
- Bal.:100 µl
Specifications
About this item
Recommended Dilutions: ELISA: 1:2000-5000; Western Blot: 1:500-2000
Type: Primary
Antigen: SOX8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat