Anti-TP53 Mouse Monoclonal Antibody [clone: DO-1]
Catalog # ORIGCF804804
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen symbol:TP53
- Clonality:Monoclonal
- Clone:DO-1
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2a
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Form:Lyophilized
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:Recombinant human wild type p53 protein expressed in E. coli.
- Purification:purified by affinity chromatography
- Size:100 μg
- Pk:100 µl
Specifications
About this item
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150
Type: Primary
Antigen: TP53
Clonality: Monoclonal
Clone: DO-1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human