Order Entry
Czech Republic
ContactUsLinkComponent
 
Anti-DUSP10 Mouse Monoclonal Antibody [clone: 2E3]
  ORIGCF806723
 :  OriGene
Anti-DUSP10 Mouse Monoclonal Antibody [clone: 2E3]
  ORIGCF806723
 :  OriGene

 

  • Antibody type:
    Primary
  • Antigen name:
    Dual specificity phosphatase 10
  • Antigen symbol:
    DUSP10
  • Clonality:
    Monoclonal
  • Clone:
    2E3
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • Isotype:
    IgG2b
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Form:
    Lyophilized
  • Amino acid number:
    180 to 482
  • Storage buffer:
    PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    –20 °C
  • Immunogen:
    Human recombinant protein fragment corresponding to amino acids 180-482 of human DUSP10(NP_009138) produced in E.coli.
  • Purification:
    purified by affinity chromatography
  • Size:
    100 μg
  • Pk:
    100 µl

 

 

Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: DUSP10
Clonality: Monoclonal
Clone: 2E3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human