Order Entry
Czech Republic
ContactUsLinkComponent
 
Anti-CSF1 Mouse Monoclonal Antibody [clone: 1F8]
  ORIGCF806609
 :  OriGene
Anti-CSF1 Mouse Monoclonal Antibody [clone: 1F8]
  ORIGCF806609
 :  OriGene

 

  • Antibody type:
    Primary
  • Antigen symbol:
    CSF1
  • Clonality:
    Monoclonal
  • Clone:
    1F8
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Form:
    Lyophilized
  • Amino acid number:
    223 to 496
  • Storage buffer:
    PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
  • Molecular weight:
    59.9 kDa
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    –20 °C
  • Immunogen:
    Human recombinant protein fragment corresponding to amino acids 223-496 of human CSF1(NP_000748) produced in E.coli.
  • Purification:
    purified by affinity chromatography
  • Size:
    100 μg
  • Pk:
    100 µl

 

 

Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: CSF1
Clonality: Monoclonal
Clone: 1F8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human