Anti-NKX2-1 Rabbit Polyclonal Antibody
Catalog # ORIGTA319719
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen name:NK2 homeobox 1
- Antigen symbol:NKX2-1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Form:Liquid
- Storage buffer:NKX2-1 Antibody is supplied in PBS containing 0,02% sodium azide.
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:NKX2-1 antibody was raised against a 16 amino acid synthetic peptide near the amino terminus of human NKX2-1.
- Purification:purified by affinity chromatography via peptide column
- Size:100 μg
- Pk:100 µG
Specifications
About this item
Recommended Dilutions: Western Blot: 1-2 ?g/ml; Immunochemistry: 2.5 ?g/ml; Immunofluorescence: 20 ?g/ml
Type: Primary
Antigen: NKX2-1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat