Anti-GSK3A Rabbit Polyclonal Antibody
Catalog # ORIGTA319232
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen name:Glycogen synthase kinase 3 alpha
- Antigen symbol:GSK3A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Form:Liquid
- Storage buffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:This affinity purified antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding aa 16-25 of human GSK3 alpha.
- Size:100 μg
- Pk:100 µG
Specifications
About this item
Recommended Dilutions: ELISA: 1:4000-1:20000; Western Blot: 1:500-1:2000
Type: Primary
Antigen: GSK3A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human