Order Entry
Czech Republic
ContactUsLinkComponent
Anti-FLT3LG Mouse Monoclonal Antibody [clone: 8D10]
Anti-FLT3LG Mouse Monoclonal Antibody [clone: 8D10]
  ORIGCF500274
 :  OriGene
Anti-FLT3LG Mouse Monoclonal Antibody [clone: 8D10]
  ORIGCF500274
 :  OriGene

 

  • Antibody type:
    Primary
  • Antigen symbol:
    FLT3LG
  • Clonality:
    Monoclonal
  • Clone:
    8D10
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Form:
    Lyophilized
  • Amino acid number:
    27 to 181
  • Storage buffer:
    PBS (pH 7,3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
  • Molecular weight:
    26.4 kDa
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    –20 °C
  • Immunogen:
    Recombinant protein expressed in E.coli corresponding to amino acids 27-181 of human FLT3LG
  • Purification:
    purified by affinity chromatography
  • Size:
    100 μg
  • Pk:
    100 µG

 

 

Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: FLT3LG
Clonality: Monoclonal
Clone: 8D10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human