Anti-GAS7 Mouse Monoclonal Antibody [clone: 9H6]
ORIGCF501873
: OriGene
- Antibody type:Primary
- Antigen symbol:GAS7
- Clonality:Monoclonal
- Clone:9H6
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Human
- Western blot:Yes
- Epitope:Full length
- Cross adsorption:No
- Form:Lyophilized
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:39.0 kDa
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:Full length human recombinant protein of human GAS7 (NP_003635) produced in HEK293T cell.
- Purification:purified by affinity chromatography
- Size:100 μg
- Pk:100 µG
Recommended Dilutions: Western Blot: 1:1000; Flow Cytometry: 1:100
Type: Primary
Antigen: GAS7
Clonality: Monoclonal
Clone: 9H6
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human