Order Entry
Czech Republic
ContactUsLinkComponent
Anti-SERPINB9 Mouse Monoclonal Antibody [clone: PI9-17]
Anti-SERPINB9 Mouse Monoclonal Antibody [clone: PI9-17]
Catalog # ORIGTA318921
Supplier:  OriGene
Anti-SERPINB9 Mouse Monoclonal Antibody [clone: PI9-17]
Catalog # ORIGTA318921
Supplier:  OriGene

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Serpin peptidase inhibitor, clade B (ovalbumin), member 9
  • Antigen symbol:
    SERPINB9
  • Clonality:
    Monoclonal
  • Clone:
    PI9-17
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Epitope:
    full length
  • Cross adsorption:
    No
  • Form:
    Liquid
  • Storage buffer:
    100 µg (0,5 mg/ml) mouse monoclonal antibody from culture supernatant diluted in phosphate-buffered saline (PBS) containing 50% glycerol, 1% BSA, and 0,02% thimerosal.
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    –20 °C
  • Immunogen:
    Recombinant human full length PI9
  • Purification:
    purified by affinity chromatography
  • Size:
    100 μg
  • Pk:
    100 µG

Specifications

About this item

Recommended Dilutions: Western Blot: 2-4 ?g/ml; Immunochemistry: 20?g/ml

Type: Primary
Antigen: SERPINB9
Clonality: Monoclonal
Clone: PI9-17
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Human