ContactUsLinkComponent
Woulff bottles
Woulff bottles
Catalog # WITG5.663.000
Supplier:  witeg Labortechnik
undefined
Woulff bottles
Catalog # WITG5.663.000
Supplier:  witeg Labortechnik
Specifications
  • Capacity:
    500 ml
  • Pk:
    1
Specifications
About this item
Glass, clear, plastic coated.
  • Clear plastic coated to protect against mechanical breakage
  • Reduces the hazard of flying glass and liquid splashes in the event of breakage
  • Implosion protection
Complete set includes 500 ml bottle with 3 necks (center neck GL 25, side necks GL 18), suction tube with safety valve, analog pressure gauge. Air-suction tube icnludes PTFE needle valve stopcock.