N,N-Dimethylformamide 99.0-100.00%, PESTINORM® for headspace gas chromatography
:68-12-2
:
:HCON(CH₃)₂
:MFCD00003284
:73.09 g/mol
:13,03269
:153 °C (1013 hPa)
:–60.5 °C
:0.949 g/cm³ (20 °C)
:Ambient
:200-679-5
:2265
:3,III
:Filtered through a 0.2 µm filter, packaged under nitrogen.
:57.5 °C (DIN 51755) (closed cup)
:01-2119475605-32
:GHS02
:GHS08
:GHS07
- Purity (GC) :min. 99.8%
- Identity (IR) :Complying
- Residue after evaporation :max. 0.0003%w/w
- Water (KF) :max. 0.05%w/w
- Color (APHA Pt-Co) :max. 10
- GC-HS (Each class 1 solvent) :max. 1µg/g
- GC-HS (Each class 2 solvent) :max. 10µg/g
- GC-HS (Each class 3 solvent) :max. 50µg/g
- Application HS-GC (standard measurement) :Passes test
- Headspace test for O.V.I's:Passes test
Frequently Bought Together