ContactUsLinkComponent
Film applicators, multi-functional
Film applicators, multi-functional
Catalog # SIMEVF1550
Supplier:  Industrial Physics
undefined
Film applicators, multi-functional
Catalog # SIMEVF1550
Supplier:  Industrial Physics
Specifications
  • Description:
    Octoplex film applicator
  • Pk:
    1
Specifications
About this item
Multi-functional film applicator (Octoplex) with 8 pre-defined thickness sides for applying paint films of 25, 50, 75, 100, 125, 150, 175, 200 µm. Film width 60 mm. The high-grade stainless steel will not be affected by acid or base elements.