Orders LinkContactUsLinkComponent
Snap vials, clean
Snap vials, clean
   K dispozici jsou možnosti udržitelnosti
Dodavatel:   EP SCIENTIFIC
Snap vials, clean
Dodavatel:  EP SCIENTIFIC
   K dispozici jsou možnosti udržitelnosti
Certifikáty
Snap vials, clean
3 Dostupných variant níže
Product Details & Documents
Clean snap VOA vials feature a snap-off septum guard that protects the septum from exposure to the general environment. The clean snap cap reduces the chance of volatile organic contaminants in the environment from passing through the septum and contaminating the sample during transport and storage. It also keeps the septum clean and free from foreign particles during handling. The clean snap cap easily peels away to provide access to the septum at the time of analysis.
  • Amber borosilicate glass vial is recommended for light sensitive samples
  • Septa permanently bonded to the cap
  • Clean snap closure features a fully integrated, snap-off septum guard that helps protect the septum from chemical and particulate contamination during handling and storage
  • Standard cap dimensions are automation friendly
  • Fully assembled
Specifications for Product Family
Specifikace
  • Catalog No:
  • EPSCSS246-0040
  • EPSCSS136-0040
  • EPSCSS236-0040
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
Specifikace
Product Family Options
Amber
Informace o produktu
  • Description
    Capacity
    Ø×H
    Colour
    Cap included
    Cap Size
    Septum
    Thickness
    Dostupnost
    Price
  • Snap vial with VOA and not certificate
    40,00 ml
    28×95 mm
    Amber
    Yes
    24-414
    Bonded
    0,125"
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 72 KS
    1 * 72 KS - 9 950,00 Kč
    9 950,00 Kč
    1 * 72 KS
       9 950,00 Kč
    Specifikace:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    EPSCSS246-0040
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
Clear
Informace o produktu
  • Description
    Capacity
    Ø×H
    Colour
    Cap included
    Cap Size
    Septum
    Thickness
    Dostupnost
    Price
  • Snap vial without certificate
    40,00 ml
    28×95 mm
    Clear
    Yes
    24-414
    Bonded
    0,125"
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 72 KS
    1 * 72 KS - 8 990,00 Kč
    8 990,00 Kč
    1 * 72 KS
       8 990,00 Kč
    Specifikace:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    EPSCSS136-0040
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • Snap vial with VOA and not certificate
    40,00 ml
    28×95 mm
    Clear
    Yes
    24-414
    Bonded
    0,125"
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 72 KS
    1 * 72 KS - 9 950,00 Kč
    9 950,00 Kč
    1 * 72 KS
       9 950,00 Kč
    Specifikace:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    EPSCSS236-0040
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact