Order Entry
Czech Republic
ContactUsLinkComponent
 
Anti-TLR7 Rabbit Polyclonal Antibody
Catalog # BSBTPB9452
Supplier:  Boster Bio
undefined
Anti-TLR7 Rabbit Polyclonal Antibody
Catalog # BSBTPB9452
Supplier:  Boster Bio

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Toll-like Receptor 7
  • Antigen symbol:
    TLR7
  • Clonality:
    Polyclonal
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Form:
    Lyophilized
  • Gene ID:
    51284
  • Antigen synonyms:
    TLR7-like
  • UniProtKB:
    Q9NYK1
  • Storage temperature:
    At -20˚C for one year. After reconstitution, at 4˚C for one month. It can also be aliquotted and stored frozen at -20˚C for a longer time. Avoid repeated freezing and thawing.
  • Immunogen:
    A synthetic peptide corresponding to a sequence at the C-terminus of human TLR7 (1020-1047aa QAHPYFWQCLKNALATDNHVAYSQVFKE), different from the related mouse sequence by two amino acids.
  • Purification:
    Immunogen affinity purified.
  • Size:
    100 µg/vial
  • Pk:
    100 µG

Specifications

About this item

Rabbit IgG polyclonal antibody for Toll-like receptor 7(TLR7) detection. Tested with WB, IHC-P, IHC-F, ICC in Human;Mouse;Rat.

TLR7 (Toll-like receptor 7) is protein that in humans is encoded by the TLR7 gene. Orthologs are found in mammals and birds. TLR7 is a member of the Toll-like receptor (TLR) family which plays a fundamental role in pathogen recognition and activation of innate immunity. TLRs are highly conserved from Drosophila to humans and share structural and functional similarities. They recognize pathogen-associated molecular patterns (PAMPs) that are expressed on infectious agents, and mediate the production of cytokines necessary for the development of effective immunity. The various TLRs exhibit different patterns of expression. This gene is predominantly expressed in lung, placenta, and spleen, and lies in close proximity to another family member, TLR8, on human chromosome X. TLR7 recognises single stranded RNA in endosomes, which is a common feature of viral genomes which are internalised by macrophages.

Add 0.2ml of distilled water will yield a concentration of 500ug/ml.

Each vial contains 5mg BSA, 0.9mg NaCl, 0.2mg Na2HPO4, 0.05mg NaN3.

WB: The detection limit for TLR7 is approximately 0.1ng/lane under reducing conditions.
Tested Species: In-house tested species with positive results.
By Heat: Boiling the paraffin sections in 10mM citrate buffer, pH6.0, for 20mins is required for the staining of formalin/paraffin sections.
Other applications have not been tested.
Optimal dilutions should be determined by end users.

Type: Primary
Antigen: VCAN
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat