Order Entry
Czech Republic
ContactUsLinkComponent
Anti-RUNX3 Rabbit Polyclonal Antibody
Anti-RUNX3 Rabbit Polyclonal Antibody
Catalog # BSBTPB9432
Supplier:  Boster Bio
undefined
Anti-RUNX3 Rabbit Polyclonal Antibody
Catalog # BSBTPB9432
Supplier:  Boster Bio

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Runt-related Transcription Factor 3
  • Antigen symbol:
    RUNX3
  • Clonality:
    Polyclonal
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    Lyophilized
  • Gene ID:
    864
  • Antigen synonyms:
    AML2|CBFA3|PEBP2aC
  • UniProtKB:
    Q13761
  • Storage temperature:
    At -20˚C for one year. After reconstitution, at 4˚C for one month. It can also be aliquotted and stored frozen at -20˚C for a longer time. Avoid repeated freezing and thawing.
  • Immunogen:
    E.coli-derived human RUNX3 recombinant protein (Position: M128-Y270). Human RUNX3 shares 93% amino acid (aa) sequence identity with mouse RUNX3.
  • Purification:
    Immunogen affinity purified.
  • Size:
    100 µg/vial
  • Pk:
    100 µG

Specifications

About this item

Rabbit IgG polyclonal antibody for Runt-related transcription factor 3(RUNX3) detection. Tested with WB, IHC-P in Human;Mouse;Rat.

Runt-related transcription factor 3, also called AML2 is a protein that in humans is encoded by the RUNX3 gene. The RUNX3 gene encodes a Runt-related transcription factor, which is part of the RUNX gene family. By fluorescence in situ hybridization, RUNX3 was assigned to human chromosome 1p36.11. RUNX3 binds to the core site of murine Leukemia virus, the core sequences in the enhancer of the polyomavirus, and also to the enhancers of the T-cell receptor genes. It may be involved in the control of cellular proliferation and/or differentiation.

Add 0.2ml of distilled water will yield a concentration of 500ug/ml.

Each vial contains 5mg BSA, 0.9mg NaCl, 0.2mg Na2HPO4, 0.05mg NaN3.

Tested Species: In-house tested species with positive results.
By Heat: Boiling the paraffin sections in 10mM citrate buffer, pH6.0, for 20mins is required for the staining of formalin/paraffin sections.
Other applications have not been tested.
Optimal dilutions should be determined by end users.

Type: Primary
Antigen: SDHA
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat