- Antibody type:Primary
- Antigen name:ataxin 1
- Antigen symbol:ATXN1
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:Lyophilized
- Gene ID:6310
- Antigen synonyms:D6S504E|SCA1|ATX1
- UniProtKB:P54253
- Storage temperature:At -20˚C for one year. After reconstitution, at 4˚C for one month. It can also be aliquotted and stored frozen at -20˚C for a longer time. Avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to a sequence at the C-terminus of human Ataxin 1 (778-808aa ESRKLEKSEDEPPLTLPKPSLIPQEVKICIE), different from the related mouse and rat sequences by one amino acid.
- Purification:Immunogen affinity purified.
- Size:100 µg/vial
- Pk:100 µG
Rabbit IgG polyclonal antibody for Ataxin-1(ATXN1) detection. Tested with WB, IHC-P in Human.
Ataxin-1 is a protein that in humans is encoded by the ATXN1 gene. The ATXN1 gene had been mapped to 6p23 by in situ hybridization. Ataxin-1 (ATXN1), a causative factor for spinocerebellar ataxia type 1 (SCA1), and the related Brother of ATXN1 (BOAT1) are human proteins involved in transcriptional repression. ATXN1 and BOAT1 might participate in several Notch-controlled developmental and pathological processes.
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Each vial contains 5mg BSA, 0.9mg NaCl, 0.2mg Na2HPO4, 0.05mg NaN3.
Tested Species: In-house tested species with positive results.
By Heat: Boiling the paraffin sections in 10mM citrate buffer, pH6.0, for 20mins is required for the staining of formalin/paraffin sections.
Other applications have not been tested.
Optimal dilutions should be determined by end users.
Type: Primary
Antigen: ATXN3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat