- Antibody type:Primary
- Antigen name:phosphatase and tensin homolog
- Antigen symbol:PTEN
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat
- Western blot:Yes
- Form:Lyophilized
- Gene ID:5728
- Antigen synonyms:CWS1|BZS|MMAC1|DEC|GLM2|MHAM|10q23del|TEP1|PTEN1
- UniProtKB:P60484
- Storage temperature:At -20˚C for one year. After reconstitution, at 4˚C for one month. It can also be aliquotted and stored frozen at -20˚C for a longer time. Avoid repeated freezing and thawing.
- Immunogen:E.coli-derived human PTEN recombinant protein (Position: E91-V403). Human PTEN shares 99.7% amino acid (aa) sequence identity with mouse PTEN.
- Purification:Immunogen affinity purified.
- Size:100 µg/vial
- Pk:100 µG
Rabbit IgG polyclonal antibody for Phosphatidylinositol 3,4,5-trisphosphate 3-phosphatase and dual-specificity protein phosphatase PTEN(PTEN) detection. Tested with WB in Human;Rat.
PTEN is also known as BZS, DEC, CWS1, GLM2, MHAM, TEP1, PTEN1. It is mapped to 10q23.3. This gene was identified as a tumor suppressor that is mutated in a large number of cancers at high frequency. The protein encoded by this gene is a phosphatidylinositol-3,4,5-trisphosphate 3-phosphatase. It contains a tensin like domain as well as a catalytic domain similar to that of the dual specificity protein tyrosine phosphatases. Unlike most of the protein tyrosine phosphatases, this protein preferentially dephosphorylates phosphoinositide substrates. The PTEN structure reveals a phosphatase domain that is similar to protein phosphatases but also has an enlarged active site important for the accommodation of the phosphoinositide substrate.
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Each vial contains 5mg BSA, 0.9mg NaCl, 0.2mg Na2HPO4, 0.05mg NaN3.
WB: The detection limit for PTEN is approximately 0.1ng/lane under reducing conditions.
Tested Species: In-house tested species with positive results.
Predicted Species: Species predicted to be fit for the product based on sequence similarities.
Other applications have not been tested.
Optimal dilutions should be determined by end users.
Type: Primary
Antigen: PTGER2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human