Zadání objednávky
Czech Republic
ContactUsLinkComponent
Anti-PPP3CA Rabbit Polyclonal Antibody
Anti-PPP3CA Rabbit Polyclonal Antibody
Catalog # BSBTPB9382
Supplier:  Boster Bio
undefined
Anti-PPP3CA Rabbit Polyclonal Antibody
Catalog # BSBTPB9382
Supplier:  Boster Bio

Specifications

  • Druh protilátky:
    Primární
  • Název antigenu:
    protein phosphatase 3, catalytic subunit, alpha isozyme
  • Symbol antigenu:
    PPP3CA
  • Klonovalita:
    Polyclonal
  • Hostitel:
    Rabbit
  • Izotyp:
    IgG
  • Reaktivita:
    Human
  • Western blot:
    Yes
  • Formulář:
    Lyophilized
  • Identifikační číslo genu:
    5530
  • Synonyma antigenu:
    CALN|CCN1|PPP2B|CNA1|CALNA1|CALNA
  • UniProtKB:
    Q08209
  • Skladovací teplota:
    At -20˚C for one year. After reconstitution, at 4˚C for one month. It can also be aliquotted and stored frozen at -20˚C for a longer time. Avoid repeated freezing and thawing.
  • Imunogen:
    A synthetic peptide corresponding to a sequence at the C-terminus of human Calcineurin A (488-521aa DAMPSDANLNSINKALTSETNGTDSNGSNSSNIQ), different from the related mouse and rat sequences by one amino acid.
  • Čištění:
    Immunogen affinity purified.
  • Rozměry protilátky:
    100 µg/vial
  • Bal.:
    100 µG

Specifications

About this item

Rabbit IgG polyclonal antibody for Serine/threonine-protein phosphatase 2B catalytic subunit alpha isoform(PPP3CA) detection. Tested with WB in Human.

Calcineurin A is also known as PPP3CA. It is mapped to 4q24. Semsarian et al. (1999) and Musaro et al. (1999) independently showed that IGF1stimulates skeletal muscle hypertrophy and a switch to glycolytic metabolism by activating calcineurin A and inducing the nuclear translocation of transcription factor NFATC1. Semsarian et al. (1999) found that hypertrophy was suppressed by the calcineurin inhibitors cyclosporin A or FK506, but not by inhibitors of the MAP kinase or phosphatidylinositol-3-OH kinase pathways. Musaro et al. (1999) showed that expression of a dominant-negative calcineurin mutant also repressed myocyte differentiation and hypertrophy. Musaro et al. (1999) demonstrated that either IGF1 or activated calcineurin induces expression of transcription factor GATA2, which accumulates in a subset of myocyte nuclei, where it associates with calcineurin and a specific dephosphorylated isoform of NFATC1.

Add 0.2ml of distilled water will yield a concentration of 500ug/ml.

Each vial contains 5mg BSA, 0.9mg NaCl, 0.2mg Na2HPO4, 0.05mg NaN3.

Tested Species: In-house tested species with positive results.
Other applications have not been tested.
Optimal dilutions should be determined by end users.

Type: Primary
Antigen: PRDX4
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat