Order Entry
Czech Republic
ContactUsLinkComponent
Anti-SELP Rabbit Polyclonal Antibody
Anti-SELP Rabbit Polyclonal Antibody
Catalog # BSBTPB9363
Supplier:  Boster Bio
undefined
Anti-SELP Rabbit Polyclonal Antibody
Catalog # BSBTPB9363
Supplier:  Boster Bio

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Selectin P
  • Antigen symbol:
    SELP
  • Clonality:
    Polyclonal
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Form:
    Lyophilized
  • Gene ID:
    6403
  • Antigen synonyms:
    CD62|PADGEM|LECAM3|GRMP|CD62P|PSEL|GMP140
  • UniProtKB:
    P16109
  • Storage temperature:
    At -20˚C for one year. After reconstitution, at 4˚C for one month. It can also be aliquotted and stored frozen at -20˚C for a longer time. Avoid repeated freezing and thawing.
  • Immunogen:
    E.coli-derived human CD62P recombinant protein (Position: W42-G271). Human CD62P shares 75% and 76% amino acid (aa) sequence identity with mouse and rat CD62P, respectively.
  • Purification:
    Immunogen affinity purified.
  • Size:
    100 µg/vial
  • Pk:
    100 µG

Specifications

About this item

Rabbit IgG polyclonal antibody for P-selectin(SELP) detection. Tested with WB, IHC-P, ELISA in Human.

CD62P is also known as SELP or P-selectin. This gene encodes a 140 kDa protein that is stored in the alpha-granules of platelets and Weibel-Palade bodies of endothelial cells. This protein redistributes to the plasma membrane during platelet activation and degranulation and mediates the interaction of activated endothelial cells or platelets with leukocytes. The membrane protein is a calcium-dependent receptor that binds to sialylated forms of Lewis blood group carbohydrate antigens on neutrophils and monocytes. Alternative splice variants may occur but are not well documented. 

Add 0.2ml of distilled water will yield a concentration of 500ug/ml.

Each vial contains 5mg BSA, 0.9mg NaCl, 0.2mg Na2HPO4, 0.05mg NaN3.

Tested Species: In-house tested species with positive results.
By Heat: Boiling the paraffin sections in 10mM citrate buffer, pH6.0, for 20mins is required for the staining of formalin/paraffin sections.
Other applications have not been tested.
Optimal dilutions should be determined by end users.

Type: Primary
Antigen: ABCG2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat