Anti-LXR alpha Rabbit Antibody
Catalog # BSBTPA1246-1
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:Liver X receptor alpha
- Antigen symbol:LXRA
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Epitope:C-Terminal
- Cross adsorption:No
- Form:Lyophilized
- Amino acid number:371 to 385
- Sequence:NVQDQLQVERLQHTY
- Storage temperature:–20 °C
- Immunogen:A synthetic peptide corresponding to a sequence at the C-terminus of human LXR alpha (371-385aa NVQDQLQVERLQHTY), identical to the related rat and mouse sequences.
- Purification:Purified by immunoaffinity chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Rabbit IgG polyclonal antibody for Oxysterols receptor LXR-alpha(NR1H3) detection. Tested with WB in Human;Mouse;Rat.
Reconstitute with distilled water.
Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.
Type: Primary
Antigen: LXR alpha
Clonality:
Clone:
Conjugation:
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat