Anti-AIF Rabbit Antibody
Catalog # BSBTPA2232
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:Apoptosis inducing factor
- Antigen symbol:AIF
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Epitope:near the C-terminus
- Cross adsorption:No
- Form:Lyophilised
- Antigen synonyms:AIF|striatal apoptosis-inducing factor|programmed cell death 8 (apoptosis-inducing factor)|apoptosis-inducing factor 1|mitochondrial|RP3-438D16.2|COXPD6|PDCD8
- Amino acid number:596 - 613
- Sequence:EQHEDLNEVAKLFNIHED
- Storage temperature:–20 °C
- Immunogen:A synthetic peptide corresponding to a sequence at the C-terminus of human AIF (596-613aa EQHEDLNEVAKLFNIHED), identical to the related rat and mouse sequences.
- Purification:purified by immunogen affinity purification
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Rabbit IgG polyclonal antibody for Apoptosis-inducing factor 1, mitochondrial(AIFM1) detection. Tested with WB, IHC-P, IHC-F, ICC in Human, Mouse, Rat.
Reconstitute with distilled water
Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.
Type: Primary
Antigen: AIF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat