Anti-Selenium Binding Protein 1 Rabbit Antibody
Catalog # BSBTPA2216
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:Selenium Binding Protein 1
- Antigen symbol:SELENBP1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Form:Lyophilised
- Antigen synonyms:Selenium binding protein1|Selenium binding protein2|56 a selenium binding protein|SP56|SBP|Selenbp2|SBP 1|Selenbp 2|Lp56|SELENBP 1|Lpsb|SP 56|SELNBP1|Selen bp2|SBP56|Selenium binding protein 2|Lp 56|SBP1|hSBP
- Amino acid number:226 - 240
- Sequence:SHLYVWDWQRHEIVQ
- Storage temperature:–20 °C
- Immunogen:A synthetic peptide corresponding to a sequence in the middle region of human Selenium Binding Protein 1 (226-240aa SHLYVWDWQRHEIVQ).
- Purification:purified by immunogen affinity purification
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Rabbit IgG polyclonal antibody for Selenium-binding protein 1(SELENBP1) detection. Tested with WB, IHC-P in Human.
Reconstitute with distilled water
Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.
Type: Primary
Antigen: SELENBP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human