Anti-Rab3C Rabbit Antibody
BSBTPA2279
- Antibody type:Primary
- Antigen name:Ras-related protein Rab-3C
- Antigen symbol:RAB3C
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Epitope:near the C-terminus
- Cross adsorption:No
- Form:Lyophilised
- Amino acid number:204 - 217
- Sequence:AITAAKQNTRLKET
- Storage temperature:–20 °C
- Immunogen:A synthetic peptide corresponding to a sequence at the C-terminus of human Rab3C (204-217aa AITAAKQNTRLKET), different from the related mouse and rat sequences by one amino acid.
- Purification:purified by immunogen affinity purification
- Size:100 µg
- Pk:100 µG
Rabbit IgG polyclonal antibody for Ras-related protein Rab-3C(RAB3C) detection. Tested with WB in Human, Mouse, Rat.
Reconstitute with distilled water
Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.
Type: Primary
Antigen: RAB3C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat