Anti-AVPR1A Rabbit Antibody
Catalog # BSBTPA2248
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:Arginine Vasopressin Receptor 1A
- Antigen symbol:AVPR1A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Form:Lyophilised
- Antigen synonyms:AVPR1A
- Amino acid number:155 - 173
- Sequence:HPLKTLQQPARRSRLMIAT
- Storage temperature:–20 °C
- Immunogen:A synthetic peptide corresponding to a sequence in the middle region of rat AVPR1A (155-173aa HPLKTLQQPARRSRLMIAT), different from the related human and mouse sequences by one amino acid.
- Purification:purified by immunogen affinity purification
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Rabbit IgG polyclonal antibody for Vasopressin V1a receptor(AVPR1A) detection. Tested with WB in Human, Mouse, Rat.
Reconstitute with distilled water
Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.
Type: Primary
Antigen: AVPR1A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat