Order Entry
Czech Republic
ContactUsLinkComponent
Anti-Integrin beta 4 Rabbit Antibody
Anti-Integrin beta 4 Rabbit Antibody
Catalog # BSBTPA1702-1
Supplier:  Boster Bio
Anti-Integrin beta 4 Rabbit Antibody
Catalog # BSBTPA1702-1
Supplier:  Boster Bio

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Integrin Beta 4
  • Antigen symbol:
    ITGB4
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Epitope:
    C-Terminal
  • Cross adsorption:
    No
  • Form:
    Lyophilized
  • Amino acid number:
    1810 to 1822
  • Sequence:
    TLSTHMDQQFFQT
  • Storage temperature:
    –20 °C
  • Immunogen:
    A synthetic peptide corresponding to a sequence at the C-terminus of human Integrin beta 4 (1810-1822aa TLSTHMDQQFFQT), different from the related mouse and rat sequences by one amino acid.
  • Purification:
    Purified by immunogen affinity purification
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Rabbit IgG polyclonal antibody for Integrin beta-4(ITGB4) detection. Tested with WB in Human;Mouse;Rat.

Reconstitute with distilled water.

Add 0.2 ml of distilled water will yield a concentration of 500 ug/ml.

Type: Primary
Antigen: Integrin beta 4
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat