ContactUsLinkComponent
One-way stopcock, Discofix®
One-way stopcock, Discofix®
Catalog # BRAU4099117
Supplier:  B. Braun
undefined
One-way stopcock, Discofix®
Catalog # BRAU4099117
Supplier:  B. Braun
Specifications
  • Colour:
    Blue
  • Description:
    One-way stopcocks
  • Pk:
    50
Specifications
About this item
Suitable for all pressure infusion systems.
  • Continuous flow channels for accurate pressure monitoring
  • Swivel lock can be moved axially or radially for fast and secure connection
  • PVC- and latex-free