ContactUsLinkComponent
Temperature indicating kit
Temperature indicating kit
Katalogové číslo 620-9129
Supplier:  KLEINFELD
Temperature indicating kit
Katalogové číslo 620-9129
Supplier:  KLEINFELD
Specifications
  • Description:
    Thermax irreversible research kit
  • Pk:
    1
Specifications
About this item
A compilation of all thermal colors and a pattern of stripes for eight temperatures in each range.
Ordering information: Content includes 13×15 ml vials with either colours with single envelope or multiple envelope, 15 ml diluent, a brush for applying and a lab-pack with 25 self-adhesive indicators.