ContactUsLinkComponent
Microfiltration assembly, with solvent pick-up adapter, ULTRA-WARE®, KIMBLE®
  SCERSP953820-1047
Microfiltration assembly, with solvent pick-up adapter, ULTRA-WARE®, KIMBLE®
  SCERSP953820-1047
  • Capacity:
    1000 ml
  • Description:
    Microfiltration assembly with solvent pick-up adapter and ground joint, 137×430 mm
  • Pk:
    1
This microfiltration system manufactured from 33 expansion, low extractable borosilicate glass eliminates pouring HPLC solvents into funnels. The solvent pickup adapter draws solvent directly from a reagent bottle through a 47 mm filter and into a filter flask.
  • Support base with 40 to 60 micron porosity glass frit
  • Integral vacuum connection above the drip tip to prevent contamination of the vacuum line with filtrate droplets
After filtration is complete, the HI-VAC valve on the solvent pickup adapter can be turned off for vacuum degassing.
Existing ULTRA-WARE® and Millipore filtration assemblies are easily converted to in-line filtration/degassing by replacing existing funnels with solvent pickup adapters. They clamp to support bases in the same way that funnels do.
 : Conforms to USP Type I and ASTM E438, Type I, Class A requirements
 : System supplied with a solvent pickup adapter, a coarse porosity fritted glass filter base, an inner joint flask and an aluminum clamp.