Order Entry
Czech Republic
ContactUsLinkComponent
DL-α-Tocopherol (Vitamin E) ≥96% (by HPLC), liquid, Sigma-Aldrich®
Irritant
DL-α-Tocopherol (Vitamin E) ≥96% (by HPLC), liquid, Sigma-Aldrich®
Supplier:  Merck
Certificates
Irritant
DL-α-Tocopherol (Vitamin E) ≥96% (by HPLC), liquid, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Appearance (Colour):
    Yellow to Very Dark Yellow and Faint, Brown to Brown
  • Appearance (Form):
    Liquid
  • Appearance (Turbidity):
    Clear
  • Infrared Spectrum:
    Conforms to Structure
  • Purity (HPLC):
    ≥96%

Specification Test Results

Frequently Bought Together