ContactUsLinkComponent
Connectors, screw caps, threaded, KIMBLE®
Connectors, screw caps, threaded, KIMBLE®
Catalog # KIML747205-2020
Supplier:  KIMBLEKONTES
undefined
Connectors, screw caps, threaded, KIMBLE®
Catalog # KIML747205-2020
Supplier:  KIMBLEKONTES
Specifications
  • Thread:
    20-400 to 20-400
  • Seq:
    03
  • Pk:
    1
Specifications
About this item
Glass filled nylon connector, PTFE seals.
  • Temperature resistance upto 200 °C