Order Entry
Czech Republic
ContactUsLinkComponent
Anti-cytokeratins 4, 5, 6, 8, 10, 13, and 18 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: C11]
Anti-cytokeratins 4, 5, 6, 8, 10, 13, and 18 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: C11]
  COBSD3-1760
Anti-cytokeratins 4, 5, 6, 8, 10, 13, and 18 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: C11]
  COBSD3-1760

 

  • Antibody type:
    Primary
  • Antigen name:
    cytokeratins 4, 5, 6, 8, 10, 13, and 18
  • Clonality:
    Monoclonal
  • Clone:
    C11
  • Conjugation:
    APC (Allophycocyanin)
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1 kappa
  • Cross adsorption:
    none
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

Type: Primary
Antigen: cytokeratins 4, 5, 6, 8, 10, 13, and 18
Clonality: monoclonal
Clone: C11
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: