Anti-VIM Mouse Monoclonal Antibody [clone: 65]
USBIV2122-01N
- Antibody type:Primary
- Antigen name:Vimentin
- Antigen symbol:VIM
- Clonality:Monoclonal
- Clone:65
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, 1% BSA, 0,1% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified
- Size:100 µg
- Pk:100 µG
Recommended dilution: Western Blot: 1:1000
Type: Primary
Antigen: VIM (vimentin)
Clonality: monoclonal
Clone: 65
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human