Anti-VEGFA Mouse Monoclonal Antibody [clone: CH-10]
Catalog # USBIV2110-11E
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Vascular endothelial growth factor A
- Antigen symbol:VEGFA
- Clonality:Monoclonal
- Clone:CH-10
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, pH 7.6, 0,09% sodium azide.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by affinity chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: VEGFA (vascular endothelial growth factor A)
Clonality: monoclonal
Clone: CH-10
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat