Anti-TNNT2 Mouse Monoclonal Antibody [clone: M322512]
Katalogové číslo USBIT8665-24F
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:troponin T type 2 (cardiac)
- Antigen symbol:TNNT2
- Clonality:Monoclonal
- Clone:M322512
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Form:liquid
- Storage buffer:PBS, pH 7,4, 0,1% sodium azide.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:250 µg
- Pk:250 µG
Specifications
About this item
Recommended dilution: Immunochemistry: 10 ?g/ml
Type: Primary
Antigen: TNNT2 (troponin T type 2 (cardiac))
Clonality: monoclonal
Clone: M322512
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human