Anti-TFRC Mouse Monoclonal Antibody [clone: 1.A.36]
Catalog # USBIT8199-14-250
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:transferrin receptor
- Antigen symbol:TFRC
- Clonality:Monoclonal
- Clone:1.A.36
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:250 µg
- Pk:250 µG
Specifications
About this item
Type: Primary
Antigen: TFRC (transferrin receptor)
Clonality: monoclonal
Clone: 1.A.36
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human