Anti-TAR DNA Binding Protein 43 Rabbit Polyclonal Antibody
Catalog # USBIT1019-47A
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:TAR DNA-binding protein 43
- Antigen symbol:TDP43
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Monkey,Chicken,Human,Xenopus laevis,Mouse
- Western blot:Yes
- Form:liquid
- Amino acid number:350 to 414
- Storage buffer:Tris-glycine, 150mM sodium chloride, 0,05% sodium azide.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Recommended dilution: Western Blot: 0.5-2?g/ml; Immunochemistry: 1:100-1:1000
Type: Primary
Antigen: TAR DNA Binding Protein 43
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Chicken, Human, Monkey, Mouse, Xenopus laevis