Anti-STAP1 Rabbit Polyclonal Antibody
Katalogové číslo USBIS7954-60B
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Signal transducing adaptor family member 1
- Antigen symbol:STAP1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Epitope:N-terminal
- Form:liquid
- Storage buffer:PBS, 0,09% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:200 µl
- Pk:200 µl
Specifications
About this item
Recommended dilution: ELISA: 1:1000; Immunochemistry: 1:10-1:50
Type: Primary
Antigen: STAP1 (signal transducing adaptor family member 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human