Anti-SSA1/Ro-52 Rabbit Polyclonal Antibody
USBIS6600-65-100
- Antibody type:Primary
- Antigen symbol:SSA1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Rodent,Human
- Form:liquid
- Antigen synonyms:Ro-52
- Storage buffer:PBS, 0,02% sodium azide, before the addition of glycerol to 40%.
- Storage temperature:Cool
- Concentration:~0,1 mg/ml
- Shipping temperature:Ice
- Purification:purified by affinity chromatography
- Size:100 µg
- Pk:100 µG
Recommended dilution: ELISA: 1:125000
Type: Primary
Antigen: SSA1/Ro-52
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rodent