Anti-Solute Carrier Family 10 Rabbit Polyclonal Antibody (Biotin)
Catalog # USBIS5329-11B-BIOT
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen symbol:Solute Carrier Family 10
- Clonality:Polyclonal
- Conjugation:Biotin
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Epitope:internal
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:200 µl
- Pk:200 µl
Specifications
About this item
Type: Primary
Antigen: Solute Carrier Family 10
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human