Anti-SH2 Domain Protein 2A Sheep Polyclonal Antibody
USBIS1010-90P
- Antibody type:Primary
- Antigen name:SH2 Domain Protein 2A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Sheep
- Isotype:IgG
- Reactivity:Rat,Mouse
- Western blot:Yes
- Form:lyophilized powder
- Amino acid number:299 to 374
- Storage buffer:PBS, 5% trehalose.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:100 µg
- Pk:100 µG
Recommended dilution: Western Blot: 1 ?g/ml
Type: Primary
Antigen: SH2 Domain Protein 2A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Mouse, Rat