Anti-SH2 domain protein 2A Sheep Polyclonal Antibody
Catalog # USBIS1010-90K
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:SH2 domain Protein 2A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Sheep
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:lyophilized powder
- Amino acid number:279 to 389
- Storage buffer:PBS, 5% Trehalose.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by affinity chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: Western Blot: 0.5 ?g/ml
Type: Primary
Antigen: SH2 domain protein 2A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human