Order Entry
Czech Republic
ContactUsLinkComponent
Anti-SERPINB9 Mouse Monoclonal Antibody [clone: Pi9-17]
Anti-SERPINB9 Mouse Monoclonal Antibody [clone: Pi9-17]
Catalog # USBIS1000-77P2
Supplier:  US Biological
Anti-SERPINB9 Mouse Monoclonal Antibody [clone: Pi9-17]
Catalog # USBIS1000-77P2
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Serpin peptidase inhibitor, clade B (ovalbumin), member 9
  • Antigen symbol:
    SERPINB9
  • Clonality:
    Monoclonal
  • Clone:
    Pi9-17
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Format:
    Supernatant
  • Form:
    liquid
  • Storage buffer:
    0,7%BSA, 0,1% sodium azide.
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    1 ml
  • Pk:
    1 mL

Specifications

About this item

Type: Primary
Antigen: SERPINB9 (serpin peptidase inhibitor, clade B (ovalbumin), member 9)
Clonality: Monoclonal
Clone: Pi9-17
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human