Anti-Semaphorin 4B Goat Polyclonal Antibody
Catalog # USBIS0668-59
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Semaphorin 4B
- Antigen symbol:SEMA4B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:lyophilized powder
- Amino acid number:39 to 712
- Storage buffer:0,2um sterile-filtered solution
- Storage temperature:Cool
- Concentration:~0,2 mg/ml
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: ELISA: 0.5-1.0 ?g/ml; Western Blot: 1.0 ?g/ml
Type: Primary
Antigen: Semaphorin 4B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human