Order Entry
Czech Republic
ContactUsLinkComponent
Anti-Secretory Component Mouse Monoclonal Antibody (Biotin) [clone: NI 194-4 (A89-39)]
Anti-Secretory Component Mouse Monoclonal Antibody (Biotin) [clone: NI 194-4 (A89-39)]
Catalog # USBIS0630-01B
Supplier:  US Biological
Anti-Secretory Component Mouse Monoclonal Antibody (Biotin) [clone: NI 194-4 (A89-39)]
Catalog # USBIS0630-01B
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    Secretory Component
  • Clonality:
    Monoclonal
  • Clone:
    NI 194-4 (A89-39)
  • Conjugation:
    Biotin
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    lyophilized powder
  • Storage buffer:
    PBS, pH 7,2. 500ul sterile ddH2O. Dilute further
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified
  • Size:
    200 µg
  • Pk:
    200 µG

Specifications

About this item

Recommended dilution: ELISA: ?1:25; Immunochemistry: ?1:20; Western Blot: ?1:50

Type: Primary
Antigen: Secretory Component
Clonality: Monoclonal
Clone: NI 194-4 (A89-39)
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human