Anti-RUNX2 Mouse Monoclonal Antibody [clone: 1D8]
Catalog # USBIR9915-12
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Runt-related Transcription Factor 2
- Antigen symbol:RUNX2
- Clonality:Monoclonal
- Clone:1D8
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG2b kappa
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Amino acid number:251 to 351
- Storage buffer:PBS, pH 7,2.
- Storage temperature:Cool
- Concentration:~0,5 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:50 µg
- Pk:50 µG
Specifications
About this item
Recommended dilution: Immunofluorescence: 10 ?g/ml; Immunochemistry: 5 ?g/ml
Type: Primary
Antigen: RUNX2 (runt-related transcription factor 2)
Clonality: monoclonal
Clone: 1D8
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human